Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Keep as concentrated solution.
Predicted reactivity: Mouse (85%), Rat (84%), Bovine (81%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Key regulator of mitochondrial calcium uniporter (MCU) required to limit calcium uptake by MCU when cytoplasmic calcium is low. MICU1 and MICU2 form a disulfide-linked heterodimer that stimulate and inhibit MCU activity, depending on the concentration of calcium. MICU2 acts as a gatekeeper of MCU that senses calcium level via its EF-hand domains: prevents channel opening at resting calcium, avoiding energy dissipation and cell-death triggering.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calcium uptake protein 2, mitochondrial; EF hand domain family A1; EF hand domain family, member A1; EF-hand domain family member A1; EF-hand domain family, member A1; EF-hand domain-containing family member A1; Smhs2 homolog
Gene Aliases: 09C01; 1110008L20Rik; 4833427E09Rik; AU041871; EFHA1; MICU2; RGD1309934; Smhs2
UniProt ID: (Human) Q8IYU8, (Rat) Q99P63, (Mouse) Q8CD10
Entrez Gene ID: (Human) 221154, (Rat) 171433, (Mouse) 68514
Molecular Function:
calcium-binding protein
calmodulin-related
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support