Product References
Comprehensive Model for Epidermal Growth Factor Receptor Ligand Binding Involving Conformational States of the Extracellular and the Kinase Domains.
Frontiers in cell and developmental biology
Hajdu T,Váradi T,Rebenku I,Kovács T,Szöllösi J,Nagy P
MA1-24226 was used in Western Blotting to developed a model explicitly accounting for conformational changes of the epidermal growth factor (EGF) receptor (EGFR) kinase and extracellular domains, their dimerizations and ligand binding to monomeric and dimeric receptor species.
Mon Sep 28 00:00:00 EDT 2020
Correlation of AR, EGFR, and HER2 Expression Levels in Prostate Cancer: Immunohistochemical Analysis and Chromogenic In Situ Hybridization.
Cancer research and treatment
Baek KH,Hong ME,Jung YY,Lee CH,Lee TJ,Park ES,Kim MK,Yoo JH,Lee SW
MA1-24226 was used in immunohistochemistry to study AR, EGFR, and HER2 protein expresion and whether the EGFR and HER2 genes are amplified in prostate cancer
Thu Mar 01 00:00:00 EST 2012
Identification of tumor epithelium and stroma in tissue microarrays using texture analysis.
Diagnostic pathology
Linder N,Konsti J,Turkki R,Rahtu E,Lundin M,Nordling S,Haglund C,Ahonen T,Pietikäinen M,Lundin J
MA1-24226 was used in immunohistochemistry to study the use of texture analysis for the automated identification of epithelium and stroma in tumor tissue microarrays
Fri Mar 02 00:00:00 EST 2012
Identification of the receptor tyrosine kinase AXL in breast cancer as a target for the human miR-34a microRNA.
Breast cancer research and treatment
Mackiewicz M,Huppi K,Pitt JJ,Dorsey TH,Ambs S,Caplen NJ
MA1-24226 was used in immunohistochemistry to identify the targets of miR-34a microRNA in human breast cancer
Tue Nov 01 00:00:00 EDT 2011
Increased NOS2 predicts poor survival in estrogen receptor-negative breast cancer patients.
The Journal of clinical investigation
Glynn SA,Boersma BJ,Dorsey TH,Yi M,Yfantis HG,Ridnour LA,Martin DN,Switzer CH,Hudson RS,Wink DA,Lee DH,Stephens RM,Ambs S
MA1-24226 was used in immunohistochemistry to investigate the prognostic significance of inducible nitric oxide synthase in estrogen receptor-negative breast cancer
Mon Nov 01 00:00:00 EDT 2010
Antigen expression of human eccrine sweat glands.
Journal of cutaneous pathology
Li HH,Zhou G,Fu XB,Zhang L
MA1-24226 was used in immunohistochemistry to identify the phenotypic markers of human eccrine sweat glands
Sun Mar 01 00:00:00 EST 2009
Differences in the tumor microenvironment between African-American and European-American breast cancer patients.
PloS one
Martin DN,Boersma BJ,Yi M,Reimers M,Howe TM,Yfantis HG,Tsai YC,Williams EH,Lee DH,Stephens RM,Weissman AM,Ambs S
MA1-24226 was used in immunohistochemistry to compare tumor microenvironments between African-American and European-American breast cancer patients
Mon Apr 13 00:00:00 EDT 2009
Establishment and characterization of a new Ewing's sarcoma cell line from a malignant pleural effusion.
Anticancer research
Schiavo R,Tullio C,La Grotteria M,Andreotti IC,Scarpati B,Romiti L,Bozzi F,Pedrazzoli P,Siena S
MA1-24226 was used in immunohistochemistry to characterise a new Ewing's sarcoma cell line derived from a malignant pleural effusion
Tue Nov 27 00:00:00 EST 2007
Tyrosine phosphorylation controls PCNA function through protein stability.
Nature cell biology
Wang SC,Nakajima Y,Yu YL,Xia W,Chen CT,Yang CC,McIntush EW,Li LY,Hawke DH,Kobayashi R,Hung MC
MA1-24226 was used in immunohistochemistry to study the role of protein phosphorylation in controlling proliferating cell nuclear antigen function
Fri Dec 01 00:00:00 EST 2006
Gemistocytic astrocytomas: histomorphology, proliferative potential and genetic alterations--a study of 32 cases.
Journal of neuro-oncology
Avninder S,Sharma MC,Deb P,Mehta VS,Karak AK,Mahapatra AK,Sarkar C
MA1-24226 was used in immunohistochemistry to investigate the clinicopathological features of gemistocytic astrocytomas
Thu Jun 01 00:00:00 EDT 2006
Higher expression of epidermal growth factor receptor is associated with extracellular matrix metalloprotease inducer in colorectal adenocarcinoma: tissue microarray analysis of immunostaining score with clinicopathological parameters.
Disease markers
Jin JS,Wu CY,Lin YF,Wang JY,Yu CP,Sheu LF,Chiang H,Tsai WC,Lee WH
MA1-24226 was used in immunohistochemistry to investigate the expression of epidermal growth factor receptor and extracellular matrix metalloprotease inducer in colorectal cancers
Thu Apr 12 00:00:00 EDT 2007
Hepatocyte growth factor-regulated tyrosine kinase substrate (HRS) interacts with PELP1 and activates MAPK.
The Journal of biological chemistry
Rayala SK,Hollander Pd,Balasenthil S,Molli PR,Bean AJ,Vadlamudi RK,Wang RA,Kumar R
MA1-24226 was used in immunohistochemistry to study the interaction of hepatocyte growth factor-regulated tyrosine kinase substrate with PELP1 and its role in MAPK activation
Fri Feb 17 00:00:00 EST 2006
Sulindac metabolites inhibit epidermal growth factor receptor activation and expression.
Journal of carcinogenesis
Pangburn HA,Kraus H,Ahnen DJ,Rice PL
MA1-24226 was used in western blot to test if sulindac metabolites block activation or expression of the epidermal growth factor receptor
Fri Sep 02 00:00:00 EDT 2005
Changes in the expression and cytological localization of betacellulin and its receptors (ErbB-1 and ErbB-4) in the trophoblasts in human placenta over the course of pregnancy.
European journal of endocrinology
Tanimura K,Nakago S,Murakoshi H,Takekida S,Moriyama T,Matsuo H,Hashimoto K,Maruo T
MA1-24226 was used in immunohistochemistry to study the expression of betacellulin and its receptors in trophoblasts in human placenta over the course of pregnancy
Thu Jul 01 00:00:00 EDT 2004
Characterization of putative stem cell phenotype in human limbal epithelia.
Stem cells (Dayton, Ohio)
Chen Z,de Paiva CS,Luo L,Kretzer FL,Pflugfelder SC,Li DQ
MA1-24226 was used in immunohistochemistry to evaluate the stem cell molecular markers in human limbal epithelia
Tue Oct 04 00:00:00 EDT 2005
Serum concentrations of growth factors in women with and without endometriosis: the action of anti-endometriosis medicines.
International immunopharmacology
Matalliotakis IM,Goumenou AG,Koumantakis GE,Neonaki MA,Koumantakis EE,Dionyssopoulou E,Athanassakis I,Vassiliadis S
MA1-24226 was used in ELISA to study the effect of anti-endometriosis medicines on serum growth factor levels in women with endometriosis
Wed Jan 01 00:00:00 EST 2003
Expression of serum human leukocyte antigen and growth factor levels in a Greek family with familial endometriosis.
Journal of the Society for Gynecologic Investigation
Matalliotakis IM,Goumenou AG,Koumantakis GE,Athanassakis I,Dionyssopoulou E,Neonaki MA,Vassiliadis S
MA1-24226 was used in ELISA to study the levels of serum leukocyte antigen and VEGF in a family with familial endometriosis
Sat Feb 01 00:00:00 EST 2003