Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MDSPCQPQPL SQALPQLPGS SSEPLEPEPG RARMGVESYL PCPLLPSYHC PGVPSEASAG SGTPRATATS TTASPLRDGF GGQDGGELRP LQSEGAAALV TKGCQRLAAQ GARPEAPKRK WAEDGGDAPS PSKRPWARQE NQEAEREGGM SCSCSSGSGE ASAGLMEEAL PSAPERLALD YIVPCMRYYG ICVKDSFLGA ALGGRVLAEV EALKRGGRLR DGQLVSQRAI PPRSIRGDQI AWVEGHEPGC RSIGALMAHV DAVIRHCAGR LGSYVINGRT KAMVACYPGN GLGYVRHVDN PHGDGRCITC IYYLNQNWDV KVHGGLLQIF PEGRPVVANI EPLFDRLLIF WSDRRNPHEV KPAYATRYAI TVWYFDAKER AAAKDKYQLA SGQKGVQVPV SQPPTPT
Prolyl hydroxylase domain proteins HIF PHD1, HIF PHD2 and HIF PHD3 (known as PHD1, PHD2 and PHD3 in rodents, respectively) can hydroxylate HIF-alpha subunits. Hypoxia-inducible factor (HIF) is a transcriptional regulator important in several aspects of oxygen homeostasis. The prolyl hydroxylases catalyze the posttranslational formation of 4-hydroxyproline in HIF-alpha proteins. HIF PHD1, which is widely expressed, with highest levels of expression in testis, functions as a cellular oxygen sensor and is important in cell growth regulation. HIF PHD1 can localize to the nucleus or the cytoplasm and is also detected in hormone responsive tissues, such as normal and cancerous mammary, ovarian and prostate epithelium. HIF PHD1 is encoded by EGLN2, which maps to chromosome 19q13. 3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Egl nine homolog 2; egl-9 family hypoxia-inducible factor 2; EIT-6; Estrogen-induced tag 6; HIF-P4H 1; HIF-PH1; HIF-prolyl hydroxylase 1; HPH-3; Hypoxia-inducible factor prolyl hydroxylase 1; P4H1; PHD1; Prolyl hydroxylase domain-containing protein 1; Prolyl hydroxylase EGLN2
Gene Aliases: EGLN2; EIT6; HIF-PH1; HIFPH1; HPH-1; HPH-3; PHD1
UniProt ID: (Human) Q96KS0
Entrez Gene ID: (Human) 112398
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support