Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATP- and membrane-binding protein that probably controls membrane reorganization/tubulation upon ATP hydrolysis. Plays a role in early endosomal transport. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EH domain containing protein MPAST2; EH domain-containing protein 4; EH-domain containing 4; Hepatocellular carcinoma-associated protein 10/11; hepatocellular carcinoma-associated protein HCA11; mPAST2; ortholog of rat pincher; PAST homolog 2; PAST homolog 4; pincher; pinocytic chaperone
Gene Aliases: 2210022F10Rik; AI197390; AI846352; AV006278; EHD4; FKSG7; HCA10; HCA11; Past2; PAST4
UniProt ID: (Human) Q9H223, (Mouse) Q9EQP2
Entrez Gene ID: (Human) 30844, (Rat) 192204, (Mouse) 98878
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support