Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Eukaryotic initiation factor 2B (EIF2B), which is necessary for protein synthesis, is a GTP exchange factor composed of five different subunits. The protein encoded by this gene is the fourth, or delta, subunit. Defects in this gene are a cause of leukoencephalopathy with vanishing white matter (VWM) and ovarioleukodystrophy. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp586J0119; eIF-2B GDP-GTP exchange factor subunit delta; eIF2B GDP-GTP exchange factor subunit delta; eukaryotic translation initiation factor 2B subunit 4 delta; eukaryotic translation initiation factor 2B, subunit 4 delta, 67kDa; translation initiation factor eIF-2b delta subunit; translation initiation factor eIF-2B subunit delta; Translation initiation factor eIF2B subunit delta
Gene Aliases: EIF-2B; EIF2B; EIF2B4; EIF2BD; EIF2Bdelta; Jgr1a
UniProt ID: (Human) Q9UI10, (Mouse) Q61749
Entrez Gene ID: (Human) 8890, (Mouse) 13667
Molecular Function: translation initiation factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support