Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DAEEDEDDGE FSDDSGADQE KDKVKMKGYN PAEVGAAGKG YLWKAAGMNM EEEEELQQNL WGLKINMEEE SESESEQSMD SEEPDSRGGS PQMDDIKVFQ NEVLGTLQRG KEENISCDNL VLEINSLKYA YNISLKEVMQ VLSHVVLEFP LQQMDSPLDS SRYCALLLPL LKAWSPVFRN YIKRAADHLE ALAAIEDFFL EHEALGISMA KVLMAFYQLE ILAEETILSW FSQRDTTDKG QQLRKNQQLQ RFIQWLK
This gene encodes one of five subunits of eukaryotic translation initiation factor 2B, a GTP exchange factor for eukaryotic initiation factor 2 and an essential regulator for protein synthesis. Mutations in this gene and the genes encoding other EIF2B subunits have been associated with leukoencephalopathy with vanishing white matter.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eIF-2B GDP-GTP exchange factor subunit epsilon; eIF2B GDP-GTP exchange factor subunit epsilon; eukaryotic translation initiation factor 2B, subunit 5 epsilon, 82kDa; initiation factor eIF-2Be; translation initiation factor eIF-2B subunit epsilon; Translation initiation factor eIF2B subunit epsilon
Gene Aliases: C81315; CACH; CLE; EIF-2B; EIF2B5; EIF2BE; EIF2Bepsilon; LVWM
UniProt ID: (Human) Q13144, (Mouse) Q8CHW4, (Rat) Q64350
Entrez Gene ID: (Human) 8893, (Mouse) 224045, (Rat) 192234
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support