Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MATTATMAT SGSARKRLLK EEDMTKVEFE TSEEVDVTPT FDTMGLREDL LRGIYAYGFE KPSAIQQRAI KQIIKGRDVI AQSQSGTGKT ATFSISVLQC L (1-100 aa encoded by BC004386)
eIF4A3 is a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. eIF4A3 is a nuclear matrix protein. Its amino acid sequence is highly similar to the amino acid sequences of the translation initiation factors eIF4AI and eIF4AII, two other members of the DEAD box protein family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent RNA helicase DDX48; ATP-dependent RNA helicase eIF4A-3; DEAD (Asp-Glu-Ala-Asp) box polypeptide 48; DEAD box protein 48; eIF-4A-III; eIF4A-III; Eukaryotic initiation factor 4A-III; Eukaryotic initiation factor 4A-like NUK-34; Eukaryotic translation initiation factor 4A isoform 3; eukaryotic translation initiation factor 4A, isoform 3; hNMP 265; NMP 265; Nuclear matrix protein 265
Gene Aliases: 2400003O03Rik; DDX48; eIF4A-III; EIF4A3; eIF4AIII; KIAA0111; mKIAA0111; MUK34; NMP265; NUK34; RCPS
UniProt ID: (Human) P38919, (Mouse) Q91VC3
Entrez Gene ID: (Human) 9775, (Mouse) 192170
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support