Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TTIDVILIN GRLKEGDTII VPGVEGPIVT QIRGLLLPPP MKELRVKNQY EKHKEVEAAQ GVKILGKDLE KTLAGLPLLV AYKEDEIPVL KDELIHELKQ TLNAIKLEEK GVYVQASTLG SLEALLEFLK TSEVPYAGIN IGPVHKKDVM KASVMLEHDP QYAVNLAFDV RIERDAQEMA DSLGVRIFSA EIIYHLFDAF TKYRQDYKKQ KQEEFKHIAV FPCKIKILPQ YIFNSRDPIV MGVTVEAGQV KQGTPMCVPS KNFVDIGIVT SIEINHKQVD VAKKGQEVCV KIEPIPGESP KMFGRHFEAT DILVSKISRQ SIDALKDWFR DEMQKSDWQL IVELKKVFEI I (871-1220 aa encoded by BC032639)
Accurate initiation of translation in eukaryotes is complex and requires many factors, some of which are composed of multiple subunits. The process is simpler in prokaryotes which have only three initiation factors (IF1, IF2, IF3). Two of these factors are conserved in eukaryotes: the homolog of IF1 is eIF1A and the homolog of IF2 is eIF5B. This gene encodes eIF5B. Factors eIF1A and eIF5B interact on the ribosome along with other initiation factors and GTP to position the initiation methionine tRNA on the start codon of the mRNA so that translation initiates accurately.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Annexin V-binding protein ABP-7; DKFZp434I036; eIF 5B; eIF-5B; Eukaryotic translation initiation factor 5B; FLJ10524; Translation initiation factor IF-2; translation initiation factor IF2
Gene Aliases: A030003E17Rik; BC018347; EIF5B; IF2; KIAA0741
UniProt ID: (Human) O60841, (Rat) A0JN31, (Mouse) Q05D44
Entrez Gene ID: (Human) 9669, (Rat) 308306, (Mouse) 226982
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support