Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: VLQELNVTVV TSLCRRSNVC TLVRGRQAGV CFGDSGSPLV CNGLIHGIAS FVRGGCASGL YPDAFAPVAQ FVNWIDSIIQ RSEDNPCPHP RDPDPASRTH
The neutrophil form of elastase is 218 amino acids long, with two asparagine-linked carbohydrate chains (see glycosylation). It is present in azurophil granules in the neutrophil cytoplasm. There appear to be two forms of neutrophil elastase, termed IIa and IIb.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bone marrow serine protease; ELA2A; Elastase; elastase 2, neutrophil; Elastase-2; Elastase2; granulocyte-derived elastase; HLE; Human leukocyte elastase; leukocyte elastase; Medullasin; Neutrophil elastase; PMN elastase; polymorphonuclear elastase
Gene Aliases: ELA2; ELANE; GE; HLE; HNE; NE; PMN-E; SCN1
UniProt ID: (Human) P08246
Entrez Gene ID: (Human) 1991
Molecular Function: serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support