Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: U87-MG.
Predicted reactivity: Mouse (92%), Rat (96%), Xenopus laevis (80%), Rhesus Monkey (99%), Bovine (94%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Acts as a GTPase-activating protein (GAP) toward guanine nucleotide exchange factors like ARL2, ARL3, ARF1 and ARF6, but not for GTPases outside the Arf family.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ELMO domain-containing protein 1; ELMO/CED-12 domain containing 1
Gene Aliases: ELMOD1
UniProt ID: (Human) Q8N336
Entrez Gene ID: (Human) 55531
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support