Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: CTFPLGWLYF QIGYMISLIA LFTNFYIQTY NKKGASRRKD HLKDHQNGSM AAVNGHTNSF SPLENNVKPR KLRKD
ELOVL5 plays a role in elongation of long-chain polyunsaturated fatty acids.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3-keto acyl-CoA synthase ELOVL5; elongation of long chain fatty acids family member 5; Elongation of very long chain fatty acids protein 5; elongation of very long chain fatty acids-like 5; ELOVL FA elongase 5; ELOVL family member 5, elongation of long chain fatty acids; ELOVL family member 5, elongation of long chain fatty acids (FEN1/Elo2, SUR4/Elo3-like, yeast); ELOVL fatty acid elongase 5; Fatty acid elongase 1; hELO1; homolog of yeast long chain polyunsaturated fatty acid elongation enzyme; homolog of yeast long chain polyunsaturated fatty acid elongation enzyme 2; rELO1; spinocerebellar ataxia 38; Very long chain 3-ketoacyl-CoA synthase 5; Very long chain 3-oxoacyl-CoA synthase 5; Very long chain fatty acid elongase 5; very-long-chain 3-oxoacyl-CoA synthase 5
Gene Aliases: 1110059L23Rik; AI747313; AU043003; dJ483K16.1; ELOVL2; ELOVL5; HELO1; PRO0530; rELO1; SCA38
UniProt ID: (Human) Q8NCG4, (Mouse) Q8BHI7, (Rat) Q920L7
Entrez Gene ID: (Human) 60481, (Mouse) 68801, (Rat) 171400
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support