Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 4
Positive Control: HeLa, HeLa nuclear, 293T, A431, HeLa, HepG2
Predicted Reactivity: Mouse (96%), Zebrafish (92%), Drosophila (88%), Yeast (84%), Xenopus laevis (92%), Chicken (91%), Rhesus Monkey (100%), Bovine (98%)
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Elongator complex protein 3 (ELP3) is a catalytic histone acetyltransferase subunit of the RNA polymerase II elongator complex, which is a component of the RNA polymerase II (Pol II) holoenzyme and is involved in transcriptional elongation. Elongator may play a role in chromatin remodeling and is involved in acetylation of histones H3 and probably H4. It may also have a methyltransferase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 2.3.1.48; elongation protein 3 homolog; Elongator complex protein 3; h KAT9; hELP3; tRNA uridine(34) acetyltransferase
Gene Aliases: ELP3; KAT9
UniProt ID: (Human) Q9H9T3
Entrez Gene ID: (Human) 55140
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support