Search Thermo Fisher Scientific
PA1-4203 detects the ETL protein in human samples.
PA1-4203 has been successfully used in Western blot procedures.
The PA1-4203 immunogen is a synthetic peptide corresponding to residues R(7) S S S N Q D R F I T N D(19) of human ETL.
ETL is a novel seven-transmembrane receptor that belongs to the secretin family of G-protein-coupled peptide hormone receptors and the EGF-TM7 subfamily of receptors. It is the first seven-transmembrane receptor containing EGF-like repeats that is developmentally regulated in the heart.
仅用于科研。不用于诊断过程。未经明确授权不得转售。
蛋白别名: Adhesion G protein-coupled receptor L4; EGF, latrophilin and seven transmembrane domain containing 1; EGF, latrophilin and seven transmembrane domain-containing protein 1; EGF-TM7-latrophilin-related protein; ETL protein; hCG_14667
基因别名: ADGRL4; ELTD1; ETL; KPG_003; UNQ202/PRO228
UniProt ID: (Human) Q9HBW9
Entrez Gene ID: (Human) 64123