Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ENANCTNTEGS YYCMCVPGFR SSSNQDRFIT NDGTVCIENV NANCHLDNVC IAANINKTLT KIRSIKEPVA LLQEVYRNSV TDLSPTDIIT YIEILAESSS LLGYKNNTIS
Binds to an epitope located within the peptide sequence CVPGFRSSSN as determined by overlapping synthetic peptides.
Highest antigen sequence identity to the following orthologs - mouse 53%, rat 55%.
ETL is a novel seven-transmembrane receptor that belongs to the secretin family of G-protein-coupled peptide hormone receptors and the EGF-TM7 subfamily of receptors. It is the first seven-transmembrane receptor containing EGF-like repeats that is developmentally regulated in the heart.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adhesion G protein-coupled receptor L4; EGF, latrophilin and seven transmembrane domain containing 1; EGF, latrophilin and seven transmembrane domain-containing protein 1; EGF-TM7-latrophilin-related protein; ETL protein; hCG_14667
Gene Aliases: ADGRL4; ELTD1; ETL; KPG_003; UNQ202/PRO228
UniProt ID: (Human) Q9HBW9
Entrez Gene ID: (Human) 64123
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support