Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
EMG1 involved in 40S ribosomal subunit biogenesis. It seems to play a role a methylation reaction in pre-rRNA processing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 18S rRNA (pseudouridine(1248)-N1)-methyltransferase; 18S rRNA (pseudouridine-N1-)-methyltransferase NEP1; 18S rRNA Psi1248 methyltransferase; EMG1 N1-specific pseudouridine methyltransferase; EMG1 nucleolar protein homolog; essential for mitotic growth 1; nucleolar essential protein 1; Nucleolar protein EMG1 homolog; Protein C2f; Ribosomal RNA small subunit methyltransferase NEP1; Ribosome biogenesis protein NEP1
Gene Aliases: C2F; EMG1; Grcc2f; NEP1
UniProt ID: (Human) Q92979
Entrez Gene ID: (Human) 10436, (Dog) 477708
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support