Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MSLLLLVVSA LHILILILLF VATLDKSWWT LPGKESLNLW YDCTWNNDTK TWACSNVSEN GWLKAVQVLM VLSLILCCLS FILFMFQLYT MRRGGLFYAT GLCQLCTSVA VFTGALIYAI HAEEILEKHP RGGSFGYCFA LAWVAFPLAL VSGIIYIHLR KRE
Probably involved in cell proliferation and cell-cell interactions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EMP-3; Epithelial membrane protein 3; Hematopoietic neural membrane protein 1; HNMP-1; Protein YMP
Gene Aliases: EMP3; YMP
UniProt ID: (Human) P54852
Entrez Gene ID: (Human) 2014
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support