Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MVVLSVPAEV TVILLDIEGT TTPIAFVKDI LFPYIEENVK EYLQTHWEEE ECQQDVSLLR KQAEEDAHLD GAVPIPAASG NGVDDLQQMI QAVVDNVCWQ MSLDRKTTAL KQLQGHMWRA AFTAGRMKAE FFADVVPAVR KWREAGMKVY IYSSGSVEAQ KLLFGHSTEG DILELVDGHF DTKIGHKVES ESYRKIADSI GCSTNNILFL TDVTREASAA EEADVHVAVV VRPGNAGLTD DEKTYYSLIT SFSELYLPSS T
Bifunctional enzyme that catalyzes the enolization of 2,3-diketo-5-methylthiopentyl-1-phosphate (DK-MTP-1-P) into the intermediate 2-hydroxy-3-keto-5-methylthiopentenyl-1-phosphate (HK-MTPenyl-1-P), which is then dephosphorylated to form the acireductone 1,2-dihydroxy-3-keto-5-methylthiopentene (DHK-MTPene).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2,3-diketo-5-methylthio-1-phosphopentane phosphatase; 2,3-diketo-5-methylthio-1-phosphopentane phosphatase {ECO:0000255|HAMAP-Rule:MF_03117}; acireductone synthase; E-1 enzyme; Enolase-phosphatase E1; enolase-phosphatase E1 {ECO:0000255|HAMAP-Rule:MF_03117}; ENOPH1; MASA homolog; MASA homolog {ECO:0000255|HAMAP-Rule:MF_03117}
Gene Aliases: 2310057D15Rik; BB183658; C81437; E1; ENOPH1; MASA; MST145; MSTP145; mtnC; RGD1309016
UniProt ID: (Human) Q9UHY7, (Mouse) Q8BGB7, (Rat) Q5PPH0
Entrez Gene ID: (Human) 58478, (Rabbit) 100346300, (Pig) 100511394, (Mouse) 67870, (Rat) 305177
Molecular Function:
phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support