Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: IGNYMWDPT TNKSFDIGVN KDSLMPLWWN GSEPLWVTLT KAKRKVYMYY WPGCEVEILG VRPTYCLEYK NVPTDINFAN AVSDALDSFK SGRADLAAIY HERIDVEGHH YGPASPQRKD ALKAVDTVLK YMTKWIQERG LQDRLNVIIF SDHGMTDIFW MDKVIELNKY ISLNDLQQVK DRGPVVSLWP APGKHSEIYN KLSTVEHMTV YEKEAIPSRF YYKKGKFVSP LTLVADEGWF ITENREMLPF WMNSTGRREG WQRGWHGYDN ELMDMRGIFL AFGPDFKSNF RAAPIRSVDV YNVMCNVVGI TPLPNNGSWS RVMCMLKGRA STAPPVWPSH CALALILLFL LA (90-440 aa encoded by BC035035)
Choline-specific glycerophosphodiester phosphodiesterase. Hydrolyzes the classical substrate for phospholipase C, p-nitrophenyl phosphorylcholine, while it does not hydrolyze the classical nucleotide phosphodiesterase substrate, p-nitrophenyl thymidine 5'-monophosphate. Hydrolyzes lysophosphatidylcholine (LPC) to form monoacylglycerol and phosphorylcholine but not lysophosphatidic acid, showing it has a lysophospholipase C activity. Has a preference for LPC with short (12:0 and 14:0) or polyunsaturated (18:2 and 20:4) fatty acids. Also hydrolyzes glycerophosphorylcholine and sphingosylphosphorylcholine efficiently.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B830047L21Rik; Choline-specific glycerophosphodiester phosphodiesterase; E-NPP 6; E-NPP6; Ectonucleotide pyrophosphatase/phosphodiesterase family member 6; glycerophosphocholine cholinephosphodiesterase; Glycerophosphocholine cholinephosphodiesterase ENPP6; GPC-Cpde; NPP-6
Gene Aliases: 4833421B01Rik; B830047L21Rik; D8Ertd514e; E-NPP 6; ENPP6; NPP6; UNQ1889/PRO4334
UniProt ID: (Human) Q6UWR7, (Mouse) Q8BGN3
Entrez Gene ID: (Human) 133121, (Mouse) 320981, (Rat) 306460
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support