Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: GVSSDSVGGF RYSERYDPEP KSKWDEEWDK NKSAFPFSDK LGELSDKIGS TIDDTISKFR RKDREDSPER CSDSDEEKKA RRGRSPKGEF KDEEETVTTK H
Binds to membranes enriched in phosphatidylinositol 4,5- bisphosphate (PtdIns(4,5)P2). May have a role in transport via clathrin-coated vesicles from the trans-Golgi network to endosomes. Stimulates clathrin assembly.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: clathrin interacting protein localized in the trans-Golgi region; Clathrin interactor 1; Clathrin-interacting protein localized in the trans-Golgi region; Clint; ENTH domain containing protein; Enthoprotin; epsin 4; Epsin-4; Epsin-related protein; EpsinR
Gene Aliases: AI642036; AW537616; C530049I24Rik; CLINT; CLINT1; ENTH; EPN4; EPNR; F630206G17Rik; KIAA0171; mKIAA0171
UniProt ID: (Human) Q14677
Entrez Gene ID: (Human) 9685, (Mouse) 216705, (Rat) 360515
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support