Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: LDIKDEIQQN GQTIYLRGTG DFDLCRETIQ PFMNKTNETQ TSLNGVYQPP IHFQNSEFYG FSEFYYCTED VLRMGGDYNA AKFTKAAKDY CATKWSILR
Hydrolyzes preferentially nucleoside 5'-diphosphates, nucleoside 5'-triphosphates are hydrolyzed only to a minor extent. The order of activity with different substrates is UDP >> GDP = CDP = TDP, AMP, ADP, ATP and UMP are not substrates. Preferred substrates for isoform 2 are CTP, UDP, CDP, GTP and GDP, while isoform 1 utilizes UTP and TTP.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ectonucleoside triphosphate diphosphohydrolase 4; golgi luminal UDPase; Golgi UDPase; guanosine-diphosphatase like protein; lysosomal apyrase-like 1; Lysosomal apyrase-like protein of 70 kDa; NTPDase 4; UDPase; Uridine-diphosphatase
Gene Aliases: ENTPD4; KIAA0392; LALP70; LAP70; LYSAL1; NTPDase-4; UDPase
UniProt ID: (Human) Q9Y227
Entrez Gene ID: (Human) 9583
Molecular Function:
oxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support