Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Positive Samples: 293T, PC-3, Mouse liver, Rat liver, Rat kidney
Immunogen sequence: GAETVQGLLE VAKDSIPRSH WKKTPVVLKA TAGLRLLPEH KAKALLFEVK EIFRKSPFLV PKGSVSIMDG SDEGILAWVT VNFLTGQLHG HRQETVGTLD L
The protein encoded by this gene is similar to E-type nucleotidases (NTPases)/ecto-ATPase/apyrases. NTPases, such as CD39, mediate catabolism of extracellular nucleotides. ENTPD5 contains 4 apyrase-conserved regions which is characteristic of NTPases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD39 antigen-like 4; CD39-like 4; Ectonucleoside triphosphate diphosphohydrolase 5; ER-UDPase; GDPase ENTPD5; Guanosine-diphosphatase ENTPD5; Inosine diphosphate phosphatase ENTPD5; NTPDase 5; Nucleoside diphosphatase; Nucleoside diphosphate phosphatase ENTPD5; Pcph proto-oncogene protein; proto-oncogene CPH; UDPase ENTPD5; Uridine-diphosphatase ENTPD5
Gene Aliases: AI196558; AI987697; CD39L4; ENTPD5; ER-UDPase; mNTPase; NTPDase-5; NTPDase5; PCPH
UniProt ID: (Human) O75356, (Rat) Q6P6S9, (Mouse) Q9WUZ9
Entrez Gene ID: (Human) 957, (Rat) 314312, (Mouse) 12499
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support