Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
ENTPD7 is a multi-pass membrane protein. It belongs to the GDA1/CD39 NTPase family. It preferentially hydrolyzes nucleoside 5'-triphosphates. The order of activity with respect to possible substrates is UTP > GTP > CTP.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ectonucleoside triphosphate diphosphohydrolase 7; lysosomal apyrase-like 2; Lysosomal apyrase-like protein 1; NTPDase 7; RP11-483F11.1
Gene Aliases: 1810012B13Rik; 1810020C02Rik; 2810003F23Rik; ENTPD7; Kiaa4066; LALP1; Lysal2
UniProt ID: (Human) Q9NQZ7, (Mouse) Q3TCT4
Entrez Gene ID: (Human) 57089, (Mouse) 93685
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support