Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MQCKVILLD GSEYTCDVEK RSRGQVLFDK VCEHLNLLEK DYFGLTYRDA ENQKNWLDPA KEIKKQVRSG AWHFSFNVKF YPPDPAQLSE DITRYYLCLQ LRDDIVSGRL PCSFVTLALL GS (191-312 aa encoded by BC008377)
Critical growth regulator in the pathogenesis of meningiomas. Its human homolog acts as a tumor suppressor and binds 14-3-3 proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 130kDa Protein 4.1B MEF cell isoform; 4.1B; 60kDa 4.1B MEF cell isoform; Band 4.1-like protein 3; DAL-1; Differentially expressed in adenocarcinoma of the lung protein 1; Erythrocyte membrane protein band 4.1-like 3; erythrocyte protein band 4.1-like 3; FLJ37633; HGNC:3380; mDAL-1
Gene Aliases: 4.1B; DAL-1; DAL1; DAL1P; Epb4.1l3; EPB41L3; KIAA0987; NBL3
UniProt ID: (Human) Q9Y2J2, (Mouse) Q9WV92
Entrez Gene ID: (Human) 23136, (Mouse) 13823, (Rat) 116724
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support