Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
This EPB41L3 antibody is useful for Western blot, where bands are seen at ~120 and 96 kDa.
It detects both isoforms A and B in mouse brain lysates.
It detects human isoform A strongly and isoform B more weakly.Suggested positive control: human and mouse brain lysate, HeLa whole cell lysate, Hela whole cell lysate, mouse brain lysate, human brain lysate.
Critical growth regulator in the pathogenesis of meningiomas. Its human homolog acts as a tumor suppressor and binds 14-3-3 proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 130kDa Protein 4.1B MEF cell isoform; 4.1B; 60kDa 4.1B MEF cell isoform; Band 4.1-like protein 3; DAL-1; Differentially expressed in adenocarcinoma of the lung protein 1; Erythrocyte membrane protein band 4.1-like 3; erythrocyte protein band 4.1-like 3; FLJ37633; HGNC:3380; mDAL-1
Gene Aliases: 4.1B; DAL-1; DAL1; DAL1P; Epb4.1l3; EPB41L3; KIAA0987; NBL3; XELAEV_18031863mg
UniProt ID: (Human) Q9Y2J2, (Mouse) Q9WV92
Entrez Gene ID: (Human) 23136, (Mouse) 13823, (Rat) 116724, (Xenopus) 398970
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support