Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
EPHA4 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. EphA4 encodes a protein that binds ephrin-A ligands.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EK8; epa4; EPH-like kinase 8; Ephrin type-A receptor 4; receptor protein-tyrosine kinase HEK8; TYRO1 protein tyrosine kinase; Tyrosine-protein kinase receptor MPK-3; Tyrosine-protein kinase receptor SEK; Tyrosine-protein kinase receptor SEK-1; Tyrosine-protein kinase TYRO1
Gene Aliases: 2900005C20Rik; AI385584; Cek8; EPHA4; HEK8; rb; RGD1560587; SEK; Sek1; TYRO1
UniProt ID: (Human) P54764, (Mouse) Q03137
Entrez Gene ID: (Human) 2043, (Rat) 316539, (Mouse) 13838
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support