Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: DTTGETSEIG WLTYPPGGWD EVSVLDDQRR LTRTFEACHV AGAPPGTGQD NWLQTHFVER RGAQRAHIRL HFSVRACSSL GVSGGTCRET FTLYYRQAEE
Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. The ephrin receptor encoded by this gene lacks the kinase activity of most receptor tyrosine kinases and binds to ephrin-B ligands.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EPB6; EphB6v; Epherin B6; Epherin receptor B6; Ephrin type-B receptor 6; HEP; human kinase-defective Eph-family receptor protein; MGC129910; MGC129911; Tyrosine-protein kinase-defective receptor EPH-6
Gene Aliases: EPHB6; HEP
UniProt ID: (Human) O15197
Entrez Gene ID: (Human) 2051
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support