Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Promo Code: P5718423 Get More for Less: Save on ELISA and More on ProcartaPlex Get 16% off ELISA kits and 26% off our ProcartaPlex multiplex assays ≤10 plex until June 30. Learn more
EPLIN is a cytoskeleton-associated protein that inhibits actin filament depolymerization and cross-links filaments in bundles.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1110021C24Rik; Epithelial protein lost in neoplasm; epithelial protein lost in neoplasm beta; LIM domain and actin-binding protein 1; sterol regulatory element binding protein 3
Gene Aliases: EPLIN; LIMA1; PP624; SREBP3
UniProt ID: (Human) Q9UHB6, (Rat) F1LR10
Entrez Gene ID: (Human) 51474, (Rat) 300228
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support