Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. EPRS is a multifunctional aminoacyl-tRNA synthetase that catalyzes the aminoacylation of glutamic acid and proline tRNA species.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bifunctional aminoacyl-tRNA synthetase; Bifunctional glutamate/proline--tRNA ligase; Cell proliferation-inducing gene 32 protein; glutamate tRNA ligase; Glutamatyl-prolyl-tRNA synthetase; glutaminyl-tRNA synthetase; proliferation-inducing gene 32 protein; proliferation-inducing protein 32; proline tRNA ligase; proline-tRNA ligase; prolyl-tRNA synthetase
Gene Aliases: EARS; EPRS; EPRS1; GLNS; GLUPRORS; PARS; PIG32; QARS; QPRS
UniProt ID: (Human) P07814
Entrez Gene ID: (Human) 2058
Molecular Function:
aminoacyl-tRNA synthetase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support