Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSLWESLGE SWMRPRSEWP REPQVPLYVP KFHSGWEPPV DVLQEAPWEV EGLASAPIEE VSPVSRQSIR NSQKHSPTSE PTPPGDALPP VSSPHTHRGC QPTPAMAKYV KILYDFTARN ANELSVLKDE VLEVLEDGRQ WWKLRSRSGQ AGYVPCNILG EARPEDAGAP FEQAGQKYWG PASPTHKLPP SFPGNKDELM QHMDEVNDEL IRKISNIRAQ PQRHFRVERS QPVSQPLTYE SGPDEVRAWL EAKAFSPRIV ENLGILTGPQ LFSLNKEELK KVCGEEGVRV YSQLTMQKAF LEKQQSGSEL EELMNKFHSM NQRRGEDS (389-715 aa encoded by BC002474)
The protein encoded by this gene has been shown to be involved in the regulation of cell proliferation, and in the crosstalk between the adiponectin signalling and insulin signalling pathways. The encoded protein binds many other proteins, including RAB5A, DCC, AKT2, PIK3CA, adiponectin receptors, and proteins of the NuRD/MeCP1 complex. This protein is found associated with endosomal membranes, but can be released by EGF and translocated to the nucleus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Epidermal growth factor receptor kinase substrate 8-like protein 2; epidermal growth factor receptor pathway substrate 8-like protein 2; Epidermal growth factor receptor pathway substrate 8-related protein 2; EPS8 related protein 2; EPS8-like 2; EPS8-like protein 2; EPS8-related protein 2
Gene Aliases: AI042819; AW545405; EPS8L2; EPS8R2; PP13181
UniProt ID: (Human) Q9H6S3, (Mouse) Q99K30
Entrez Gene ID: (Human) 64787, (Mouse) 98845
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support