Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: CPMSPEFYRE YVAIKTKKRI LLYTMNPNKF RACQFLIKFH ERRNDKIIVF ADNVFALKEY AIRLNKPYIY GPTSQGERMQ ILQNFKHNPK INTIFISKVG DTSFDLPEAN VLIQISSHGG SRRQEAQRLG RVLRAKKGMV AEEYNAFFYS LVSQDTQEMA YSTKRQRFLV DQGYSFKVIT KLAGMEEEDL AFSTKEEQQQ LLQKVLAATD LDAEEEVVAG EFGSRSSQAS RRFGTMSSMS GADDTVYMEY HSSRSKAPSK HVHPLFKRFR K
The human XPB DNA helicase is a subunit of the DNA repair/basal transcription factor TFIIH, and is involved in early steps of the nucleotide excision repair (NER) pathway. Two distinct clinical phenotypes, xeroderma pigmentosum associated with Cockayne's syndrome (XP/CS) and trichothiodystrophy (TTD), can be due to mutations in the XPB gene.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Basic transcription factor 2 89 kDa subunit; BTF2 p89; BTF2-p89; DNA excision repair protein ERCC-3; DNA repair helicase; DNA repair protein complementing XP-B cells; excision repair 3; excision repair cross-complementation group 3; excision repair cross-complementing rodent repair deficiency, complementation group 3; General transcription and DNA repair factor IIH helicase subunit XPB; TFIIH 89 kDa subunit; TFIIH basal transcription factor complex 89 kDa subunit; TFIIH basal transcription factor complex helicase XPB subunit; TFIIH p89; TFIIH subunit XPB; Xeroderma pigmentosum group B-complementing protein; xeroderma pigmentosum, complementation group B
Gene Aliases: BTF2; BTF2 p89; Ercc-3; ERCC3; GTF2H; RAD25; TFIIH; TTD2; XPB; XPBC
UniProt ID: (Human) P19447, (Mouse) P49135, (Rat) Q4G005
Entrez Gene ID: (Human) 2071, (Mouse) 13872, (Rat) 291703
Molecular Function: DNA metabolism protein DNA helicase nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support