Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: LGRGSVSDCS DGTSELEEPL GEDPRARPPG PPDLGAFRGP PLARLPHDPG VFRVYPRPRG GPEPLSPFPV SPLAGPGSLL PPQLSPALPM TPTHLAYTPS PTLSPMYPSG
Members of the ETS family of transcription factors, such as ERF, regulate cell proliferation and differentiation. They share a highly conserved DNA-binding domain, the ETS domain, that recognizes the sequence GGAA/T (de Castro et al., 1997 [PubMed 9192842]). For further information on ETS transcription factors, see ETS1 (MIM 164720).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Est2 repressor factor; ETS domain-containing transcription factor ERF; Ets2 repressor factor; PE-2
Gene Aliases: CRS4; ERF; PE-2; PE2
UniProt ID: (Human) P50548, (Mouse) P70459
Entrez Gene ID: (Human) 2077, (Mouse) 13875
Molecular Function:
winged helix/forkhead transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support