Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
The MW of this protein is 32 kDa, and this antibody specifically recognizes the 32 kDa protein.
Immunogen sequence: TTEVVNELY VDDPDKDSGG KIDVSLNISL PNLHCELVGL DIQDEMGRHE VGHIDNSMKI PLNNGAGCRF EGQFSINKVP GNFHVSTHSA TAQPQNPDMT HVIHKLSFGD TLQVQNIHGA FNALGGADRL TSNPLASHDY ILKIVPTVYE DKSGKQRYSY QYTVANKEYV AYSHTGRIIP AIWFRYDLSP ITVKYTERRQ PLYRFITTIC AIIGGTFTVA GILDSCIFTA SEAWKKIQLG KMH (49-290 aa encoded by BC012766)
The function of ERGIC1 remains largely unknown. ERGIC1 was highly expressed in most primary prostate tumors. It is an intriguing potential drug target especially for the ERG oncogene expressing tumors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: endoplasmic reticulum-golgi intermediate compartment (ERGIC) 1; endoplasmic reticulum-golgi intermediate compartment 32 kDa protein; Endoplasmic reticulum-Golgi intermediate compartment protein 1; ER-Golgi intermediate compartment 32 kDa protein; ERGIC-32; mKIAA1181 protein-like
Gene Aliases: 1200007D18Rik; ERGIC-32; ERGIC1; ERGIC32; HT034; KIAA1181; maa-136; NET24
UniProt ID: (Human) Q969X5, (Mouse) Q9DC16
Entrez Gene ID: (Human) 57222, (Rat) 287177, (Mouse) 67458
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support