Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The function of ERGIC1 remains largely unknown. ERGIC1 was highly expressed in most primary prostate tumors. It is an intriguing potential drug target especially for the ERG oncogene expressing tumors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: endoplasmic reticulum-golgi intermediate compartment (ERGIC) 1; endoplasmic reticulum-golgi intermediate compartment 32 kDa protein; Endoplasmic reticulum-Golgi intermediate compartment protein 1; ER-Golgi intermediate compartment 32 kDa protein; ERGIC-32
Gene Aliases: ERGIC-32; ERGIC1; ERGIC32; HT034; KIAA1181; NET24
UniProt ID: (Human) Q969X5
Entrez Gene ID: (Human) 57222
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support