Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PLDTVTFYK VIPKSKFVLV KFDTQYPYGE KQDEFKRLAE NSASSDDLLV AEVGISDYGD KLNMELSEKY KLDKESYPVF YLFRDGDFEN PVPYTGAVKV GAIQRWLKGQ GVYLGMPGCL PVYDALAGEF IRASGVEARQ ALLKQGQDNL SSVKETQKKW AEQYLKIMGK ILDQGEDFPA SEMTRIARLI EKNKMSDGKK EELQKSLNIL TAFQKKGAEK EEL (40-261 aa encoded by BC101495)
This gene encodes a reticuloplasmin, a protein which resides in the lumen of the endoplasmic reticulum. The protein shows sequence similarity to the protein disulfide isomerase family. However, it lacks the thioredoxin motif characteristic of this family, suggesting that this protein does not function as a disulfide isomerase. The protein dimerizes and is thought to play a role in the processing of secretory proteins within the ER. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: endoplasmic reticulum lumenal protein ERp28; endoplasmic reticulum protein ERp29; Endoplasmic reticulum resident protein 28; Endoplasmic reticulum resident protein 29; Endoplasmic reticulum resident protein 31; endoplasmic retuclum protein 29; epididymis secretory protein Li 107; ERp28; ERp29; ERp31; protein disulfide isomerase family A, member 9
Gene Aliases: 1200015M03Rik; 2810446M09Rik; AW209030; C12orf8; ERP28; ERP29; ERp31; HEL-S-107; PDI-DB; PDIA9
UniProt ID: (Human) P30040, (Rat) P52555, (Mouse) P57759
Entrez Gene ID: (Human) 10961, (Rat) 117030, (Mouse) 67397
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support