Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 4
Immunogen sequence: GNGVYKGFPP KWSGRCGLGY LVPSLTRYLT LNASQITNLR SFIHKVTPHR CTQGDTDNPP LYCNPKDNST IRALFPSLGT YDLEKAILNI SKAMEQEFSA TKQTLEAHQS KVSSLASASR KDHVLDIPTT QRQTACGTVG K
Highest antigen sequence identity to the following orthologs: Mouse - 23%, Rat - 20%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endogenous retroviral envelope protein HEMO; Endogenous retrovirus group MER34 member 1 Env polyprotein; endogenous retrovirus group MER34, member 1; HEMO; HERV-MER_4q12 provirus ancestral Env polyprotein; Human endogenous MER34 (medium-reiteration-frequency-family-34) open reading frame; Human endogenous MER34 ORF; LP9056 protein
Gene Aliases: ERVMER34-1; HEMO; LP9056
UniProt ID: (Human) Q9H9K5
Entrez Gene ID: (Human) 100288413
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support