Invitrogen antibodies—maximize lab budgets with advanced validation & coverage

Shop antibodies

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • ESAM Antibodies

            Invitrogen

            ESAM Monoclonal Antibody (1G8), PE, eBioscience™

            1 Published Figure
            4 References
            View all (26) ESAM antibodies

            Promotions

            View available promotion(s)

            Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more

            Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more

            Datasheet
            Protocols
            Questions & Answers
            Catalog # 12-5852-82
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            226.95
            Special offer
            Online exclusive
            267.00
            Save 40.05 (15%)
            100 µg
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite ESAM Monoclonal Antibody (1G8), PE, eBioscience™

            • Antibody Testing Data (1)
            • Published Figures (1)
            Application
            ESAM Antibody in Flow Cytometry (Flow)
            Group 53 Created with Sketch.

            FIGURE: 1 / 2

            ESAM Antibody (12-5852-82) in Flow

            Staining of bEnd.3 mouse endothelioma cell line with 0.125 µg of Rat IgG2a kappa Isotype Control PE (Product # 12-4321-80) (open histogram) or 0.125 µg of Anti-Mouse ESAM PE (filled histogram). Total viable cells were used for analysis.
            View Product

            ESAM Antibody in Flow Cytometry (Flow)
            ESAM Antibody in Flow Cytometry (Flow)

            Product Details

            12-5852-82

            Applications
            Tested Dilution
            Publications

            Flow Cytometry (Flow)

            0.25 µg/test
            View 4 publications 4 publications
            Product Specifications

            Species Reactivity

            Mouse

            Published species

            Mouse

            Host/Isotype

            Rat / IgG2a, kappa

            Recommended Isotype Control

            Rat IgG2a kappa Isotype Control (eBR2a), PE, eBioscience™

            Class

            Monoclonal

            Type

            Antibody

            Clone

            1G8

            Conjugate

            PE

            Excitation/Emission Max

            565/576 nm View spectra spectra

            Form

            Liquid

            Concentration

            0.2 mg/mL

            Purification

            Affinity chromatography

            Storage buffer

            PBS, pH 7.2

            Contains

            0.09% sodium azide

            Storage conditions

            4°C, store in dark, DO NOT FREEZE!

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_891537

            Product Specific Information

            Description: The 1G8 monoclonal antibody reacts with mouse ESAM (endothelial cell-selective adhesion molecule). ESAM is a 55-kDa membrane protein composed of 2 extracellular Ig domains, a single transmembrane domain, and a 120 amino acid cytoplasmic domain. ESAM is related to members of the junctional adhesion molecule (JAM) family, and is expressed on endothelial cells and activated platelets. ESAM at endothelial tight junctions plays a role in the regulation of vascular permeability, in response to agents such as VEGF or extravasating neutrophils.

            Applications Reported: This 1G8 antibody has been reported for use in flow cytometric analysis.

            Applications Tested: This 1G8 antibody has been tested by flow cytometric analysis. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

            Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.

            Filtration: 0.2 µm post-manufacturing filtered.

            Target Information

            ESAM is a 55-kDa membrane protein composed of 2 extracellular Ig domains, a single transmembrane domain, and a 120 amino acid cytoplasmic domain. ESAM is related to members of the junctional adhesion molecule (JAM) family, and is expressed on endothelial cells and activated platelets. ESAM at endothelial tight junctions plays a role in the regulation of vascular permeability, in response to agents such as VEGF or extravasating neutrophils.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            How to build a flow cytometry experiment with Invitrogen Flow Cytometry Panel Builder
            The Invitrogen Flow Cytometry Panel Builder webtool helps you chose antibody clones, fluorophores, fluorescent proteins, and reagents to design your experiment.
            Video Player is loading.
            Current Time 0:00
            /
            Duration 0:00
            Loaded: 0%
            0:00
            Stream Type LIVE
            Remaining Time -0:00
             
            1x
            • 2x
            • 1.75x
            • 1.5x
            • 1.25x
            • 1x, selected
            • 0.75x
            • 0.5x
            • Chapters
            • descriptions off, selected
            • captions settings, opens captions settings dialog
            • captions off, selected

              This is a modal window.

              Beginning of dialog window. Escape will cancel and close the window.

              Text
              Text Background
              Caption Area Background
              Font Size
              Text Edge Style
              Font Family

              End of dialog window.

              This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.

              How to use the Panel Builder

              Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

              Bioinformatics

              Protein Aliases: Endothelial cell-selective adhesion molecule

              View more View less

              Gene Aliases: 2310008D05Rik; Esam; Esam1; W117m

              View more View less

              UniProt ID: (Mouse) Q925F2

              View more View less

              Entrez Gene ID: (Mouse) 69524

              View more View less

              Process(es)

              cell adhesion homophilic cell adhesion via plasma membrane adhesion molecules single organismal cell-cell adhesion calcium-independent cell-cell adhesion via plasma membrane cell-adhesion molecules
              It has to be done as per old AB suggested Products section.
              guarantee_icon

              Performance Guarantee

              If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

              Learn more
              help_icon

              We're here to help

              Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

              Contact tech support
              Ordering Plus Icon Minus Icon
              • Order Status
              • Order Help
              • Quick Order
              • Supply Center
              • eProcurement
              Support Plus Icon Minus Icon
              • Help and Support
              • Contact Us
              • Technical Support Centers
              • Documents and Certificates
              • Report a Site Issue
              Resources Plus Icon Minus Icon
              • Learning Centers
              • Promotions
              • Events and Webinars
              • Social Media
              About Thermo Fisher Plus Icon Minus Icon
              • About Us
              • Careers
              • Investors
              • News
              • Social Responsibility
              • Trademarks
              • Consumer Health Data Privacy Policy
              Our Portfolio Plus Icon Minus Icon
              • Thermo Scientific
              • Applied Biosystems
              • Invitrogen
              • Gibco
              • Ion Torrent
              • Fisher Scientific
              • Unity Lab Services
              • Patheon
              • PPD
              • Terms & Conditions
              • Privacy Information Center
              • Price & Freight Policy
              • Cookie Preferences

                Your choices regarding cookies on this site

                We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
              © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
              united states flag icon
              United States

              Your items have has been added!


              Host server : magellan-search-green-56756dfb8b-mgmkt:80/100.66.75.22:80.
              git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
              git-url: https://github.com/thermofisher/magellan-search
              git-branch: release/2.31.1-2025.06.25-1.0