Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: WGKKAFSGYL GTDQSKWKAY DATHLVKSYP GSQLDILIDQ GKDDQFLLDG QLLPDNFIAA CTEKKIPVVF RLQEGYDHSY YFIATFITDH IRHHAKYLN
This gene encodes a serine hydrolase that belongs to the esterase D family. The encoded enzyme is active toward numerous substrates including O-acetylated sialic acids, and it may be involved in the recycling of sialic acids. This gene is used as a genetic marker for retinoblastoma and Wilson's disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: esterase 10; Esterase D; esterase D/formylglutathione hydrolase; FGH; Methylumbelliferyl-acetate deacetylase; S-formylglutathione hydrolase; testicular tissue protein Li 66
Gene Aliases: ESD; FGH
UniProt ID: (Human) P10768
Entrez Gene ID: (Human) 2098
Molecular Function:
protease
serine protease
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support