Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
May play a role as calcium-regulated intrinsic membrane protein. Target protein is an enriched in adipocyte plasma membranes and high density microsome fractions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E-Syt1; extended synaptotagmin like protein 1; extended synaptotagmin protein 1; Extended synaptotagmin-1; extended synaptotagmin-like protein 1; family with sequence similarity 62 (C2 domain containing), member A; membrane bound C2 domain containing protein; Membrane-bound C2 domain-containing protein; vp115
Gene Aliases: ESYT1; FAM62A; KIAA0747; MBC2
UniProt ID: (Human) Q9BSJ8, (Rat) Q9Z1X1
Entrez Gene ID: (Human) 23344, (Rat) 29579
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support