Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MISLIIPPK DQISRVAKML ADEFGTASNI KSRVNRLSVL GAITSVQQRL KLYNKVPPNG LVVYCGTIVT EEGKEKKVNI DFEPFKPINT SLYLCDNKFH TEALTALLSD DSKFGFIVID GSGALFGTLQ GNTREVLHKF TVDLPKKHGR GGQSALRFAR LRMEKRHNYV RKVAETAVQL FISGDKVNVA GLVLAGSADF KTELSQSDMF DQRLQSKVLK LVDISYGGEN GFNQAIELST EVLSNVKFIQ EKKLIGRYFD EISQDTGKYC FGVEDTLKAL EMGAVEILIV YENLDIMRYV LHCQGTEEEK ILYLTPEQEK DKSHFTDKET GQEHELIESM PLLEWFANNY KKFGATLEIV TDKSQEGSQF VKGFGGIGGI LRYRVDFQGM EYQGGDDEFF DLDDY (1-404 aa encoded by BC014269)
Termination of protein biosynthesis and release of the nascent polypeptide chain are signaled by the presence of an in-frame stop codon at the aminoacyl site of the ribosome. The process of translation termination is universal and is mediated by protein release factors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eRF1; Eukaryotic peptide chain release factor subunit 1; Eukaryotic release factor 1; polypeptide chain release factor 1; Protein Cl1; sup45 (yeast omnipotent suppressor 45) homolog-like 1; TB3-1
Gene Aliases: AI463371; D5S1995; D6Ertd109e; ERF; ERF1; ETF1; RF1; SUP45L1; TB3-1
UniProt ID: (Human) P62495, (Mouse) Q8BWY3, (Rat) Q5U2Q7
Entrez Gene ID: (Human) 2107, (Mouse) 225363, (Rat) 307503
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support