Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Aspire Free Products
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • ETFDH Antibodies

            Proteintech

            ETFDH Polyclonal Antibody

            12 References
            View all (7) ETFDH antibodies
            Datasheet
            Protocols
            Questions & Answers
            Tech Support
            Datasheet
            Protocols
            Questions & Answers
            Tech Support

            Cite ETFDH Polyclonal Antibody

            • Antibody Testing Data (16)
            Application
            ETFDH Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Group 53 Created with Sketch.

            FIGURE: 1 / 16

            ETFDH Antibody (11109-1-AP) in IHC (P)

            Immunohistochemical analysis of paraffin-embedded human liver cancer tissue slide using (Product # 11109-1-AP) (ETFDH antibody) at dilution of 1:200 (under 10x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0).
            View Product

            ETFDH Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            ETFDH Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            ETFDH Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            ETFDH Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            ETFDH Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            ETFDH Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            ETFDH Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            ETFDH Antibody in Western Blot (WB)
            ETFDH Antibody in Western Blot (WB)
            ETFDH Antibody in Western Blot (WB)
            ETFDH Antibody in Western Blot (WB)
            ETFDH Antibody in Western Blot (WB)
            ETFDH Antibody in Western Blot (WB)
            ETFDH Antibody in Western Blot (WB)
            ETFDH Antibody in Western Blot (WB)
            ETFDH Antibody in Immunoprecipitation (IP)

            Product Details

            11109-1-AP

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:2,000
            View 7 publications 7 publications

            Immunohistochemistry (IHC)

            -
            View 1 publication 1 publication

            Immunohistochemistry (Paraffin) (IHC (P))

            1:20-1:200
            -

            Immunocytochemistry (ICC/IF)

            -
            View 2 publications 2 publications

            Immunoprecipitation (IP)

            0.5-4.0 µg
            -

            Miscellaneous PubMed (Misc)

            -
            View 1 publication 1 publication

            Not applicable (N/A)

            -
            View 1 publication 1 publication
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Published species

            Human, Mouse

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            ETFDH Fusion Protein Ag1568 (269-617 aa encoded by BC011890)
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.21 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.3, with 50% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            -20°C

            Shipping conditions

            Wet ice (domestic); Dry ice (international)

            Product Specific Information

            This antibody is specific to ETFDH.

            Immunogen sequence: QTYGIGLKE LWVIDEKNWK PGRVDHTVGW PLDRHTYGGS FLYHLNEGEP LVALGLVVGL DYQNPYLSPF REFQRWKHHP SIRPTLEGGK RIAYGARALN EGGFQSIPKL TFPGGLLIGC SPGFMNVPKI KGTHTAMKSG ILAAESIFNQ LTSENLQSKT IGLHVTEYED NLKNSWVWKE LYSVRNIRPS CHGVLGVYGG MIYTGIFYWI LRGMEPWTLK HKGSDFERLK PAKDCTPIEY PKPDGQISFD LLSSVALSGT NHEHDQPAHL TLRDDSIPVN RNLSIYDGPE QRFCPAGVYE FVPVEQGDGF RLQINAQNCV HCKTCDIKDP SQNINWVVPE GGGGPAYNGM (269-617 aa encoded by BC011890)

            Target Information

            Electron-transferring-flavoprotein dehydrogenase in the inner mitochondrial membrane accepts electrons from electron-transfer flavoprotein which is located in the mitochondrial matrix and reduces ubiquinone in the mitochondrial membrane. The protein is synthesized as a 67-kDa precursor which is targeted to mitochondria and processed in a single step to a 64-kDa mature form located in the mitochondrial membrane. Deficiency in electron-transferring-flavoprotein dehydrogenase have been demonstrated in some patients with type II glutaricacidemia.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: Electron transfer flavoprotein-ubiquinone oxidoreductase, mitochondrial; electron transferring flavoprotein dehydrogenase; Electron-transferring-flavoprotein dehydrogenase; ETF dehydrogenase; ETF-QO; ETF-ubiquinone oxidoreductase

            View more View less

            Gene Aliases: 0610010I20Rik; AV001013; ETFDH; ETFQO; MADD

            View more View less

            UniProt ID: (Human) Q16134, (Mouse) Q921G7

            View more View less

            Entrez Gene ID: (Human) 2110, (Rat) 295143, (Mouse) 66841

            View more View less

            Function(s)

            electron-transferring-flavoprotein dehydrogenase activity electron carrier activity oxidoreductase activity oxidoreductase activity, oxidizing metal ions with flavin as acceptor metal ion binding quinone binding ubiquinone binding flavin adenine dinucleotide binding 4 iron, 4 sulfur cluster binding iron-sulfur cluster binding oxidoreductase

            Process(es)

            response to oxidative stress electron transport chain respiratory electron transport chain fatty acid beta-oxidation using acyl-CoA dehydrogenase biological_process oxidation-reduction process transport
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us About Us
            • Careers Careers
            • Investors Investors
            • News News
            • Social Responsibility Social Responsibility
            • Trademarks
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            Spain flag icon
            Spain

            TEST

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-8lg2n:80/100.66.79.248:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0