Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Cow: 85%; Mouse: 100%; Rat: 92%
Binds to DNA sequences containing the consensus pentanucleotide 5'-CGGA[AT]-3'.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ets related protein 71; ETS translocation variant 2; ets variant gene 2; Ets-related protein 71
Gene Aliases: Er71; Etsrp71; Etv2
UniProt ID: (Mouse) P41163
Entrez Gene ID: (Mouse) 14008
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support