Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MASPTLSPDS SSQEALSAPT CSPTSDSENL SPDELELLAK LEEQNRLLEA DSKSMRSMNG SRRNSGSSLV SSSSASSNLS HLEEDTWILW GRIANEWEEW RRRKEKLLKE LIRKGIPHHF RAIVWQLLCS ATDMPVKNQY SELLKMSSPC EKLIRRDIAR TYPEHEFFKG QDSLGQEVLF NVMKAYSLVD REVGYCQGSA FIVGLLLMQM PEEEAFCVFV RLMQEYRLRE LFKPSMAELG LCIYQFEYML QEQLPDLNTH FRSQSFHTSM YASSWFLTLF LTTFPLPVAT RVFDIFMYEG LEIVFRVGLA LLQVNQAELM QLDMEGMSQY FQRVIPHQFD SCPDKLVLKA YQVKYNPKKM KRLEKEYAAM KSKEMEEQIE IKRLRTENRL LKQRIETLEK GQVTRAQEAE ENYVIKRELA VVRQQCSSAA EDLQKAQSTI RQLQEQQENP RLTEDFVSHL ETELEQSRLR ETETLGALRE MQDKVLDMEK RNSSLPDENN VAQLQEELKA LKVREGQAVA STRELKLQLQ ELSDTWQAHL ARGGRWKESP RKLVVGELQD ELMSVRLREA QALAEGRELR QRVVELETQD HIHRNLLNRV EAERAALQEK LQYLAAQNKG LQTQLSESRR KQAEAECKSK EEVMAVRLRE ADSMAAVAEM RQRIAELEIQ REEGRIQGQL NHSDSSQYIR ELKDQIEELK AEVRLLKGPP PFEDPLAFDG LSLARHLDED SLPSSDEELL GVGVGAALQD ALYPLSPRDA RFFRRLERPA KDSEGSSDSD ADELAAPYSQ GLDN
Functions as a GTPase-activating protein (GAP) with a broad specificity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ecotropic viral integration site 5-like protein; EVI5-like protein
Gene Aliases: EVI5L
UniProt ID: (Human) Q96CN4
Entrez Gene ID: (Human) 115704
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support