Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: ESRKDMVVFL DGGQLGTLVG KRVSNLSEAV GSPLPEPPEK MVPRGCLSPR AVPPATRERG GGGPEEEPVD GLAGSAAGPG AEPQVAGAAM LGPGPPAPSV DSLSGQGQP
EVX1 is a member of the vertebrate eve-related homeo box family. This protein acts as a transcriptional repressor. A similar protein in mice is critical for early embryogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eve, even-skipped homeo box homolog 1; eve, even-skipped homeobox homolog 1; even-skipped homeo box 1 (homolog of Drosophila); EVX-1; Homeobox even-skipped homolog protein 1
Gene Aliases: EVX-1; EVX1
UniProt ID: (Human) P49640
Entrez Gene ID: (Human) 2128
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support