Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
This antibody can recognize all these three isoforms.
Immunogen sequence: SDCLSHQYG ILLNNVFDTQ VADVLQFSME TGGYLPNCIT TLQESLIKHL QVAPKYLSFL EKRQKLIQEN PEVWFIRPVS PSLLKILALE ATYLLPLRLA LLDEMMSDLT TLVDGYLNTY REGSADRLGG TEPTCMELPE ELLQLKDFQK QRREKAAREY RVNAQGLLIR TVLQPKKLVT ETAGKEEKVK GFLFGKNFRI DKAPSFTSQD FHGDVNLLKE ESLNKQATNP QHLPPTEEGE TSEDSSNKLI CTKSKGSEDQ RITQKEHFMT PKHEFQASLS LKEETEQLLM VENKEDLKCT KQAVSMSSFP QETRVSPSDT FYPIRKTVVS TLPPCPALEK IDSWISPFLN LP (164-514 aa encoded by BC030628)
EXD1 is an RNA-binding component of the PET complex, a multiprotein complex required for the processing of piRNAs during spermatogenesis. The piRNA metabolic process mediates the repression of transposable elements during meiosis by forming complexes composed of piRNAs and Piwi proteins and governs the methylation and subsequent repression of transposable elements, preventing their mobilization, which is essential for the germline integrity. The PET complex is required during the secondary piRNAs metabolic process for the PIWIL2 slicing-triggered loading of PIWIL4 piRNAs. In the PET complex, EXD1 probably acts as an RNA adapter. EXD1 is an inactive exonuclease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Exonuclease 3'-5' domain-containing protein 1; exonuclease 3'-5' domain-like 1; Exonuclease 3'-5' domain-like-containing protein 1; Inactive exonuclease EXD1; mExd1; piRNA biogenesis protein EXD1; Vexonuclease 3'-5' domain-like 1
Gene Aliases: 4932702D22Rik; EXD1; EXDL1; RGD1561009
UniProt ID: (Human) Q8NHP7, (Mouse) Q8CDF7
Entrez Gene ID: (Human) 161829, (Rat) 311334, (Mouse) 241624
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support