Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AVVVPGTRS RFFCSSDSTD CVSNKVSIQP LDETAVTDKE NNLHESEYGD QEGKRLVDTD VARNSSDDIP NNHIPGDHIP DKATVFTDEE SYSFESSKFT RTISPPTLGT LRSCFSWSGG LGDFSRTPSP SPSTALQQFR RKSDSPTSLP ENNMSDVSQL KSEESSDDES HPLREGACSS QSQESGEFSL QSSNASKLSQ CSSKDSDSEE SDCNIKLLDS QSDQTSKLCL SHFSKKDTPL RNKVPGLYKS SSADSLSTTK IKPLGPARAS GLSKKPASIQ KRKHHNAENK PGLQIKLNEL WKNFGFKKDS EKLPPCKKPL SPVRDNIQLT PEAEEDIFNK PECGRVQRAI FQ (496-846 aa encoded by BC007491)
Substrate-recognition component of the SCF (SKP1-CUL1-F-box protein)-type E3 ubiquitin ligase complex. Required for muscle atrophy following denervation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Exonuclease 1; Exonuclease I; hExo1; hExoI; mExo1; rad2 nuclease family member, homolog of S. cerevisiae exonuclease 1
Gene Aliases: 5730442G03Rik; EXO1; EXOI; HEX1; hExoI; Msa
UniProt ID: (Human) Q9UQ84, (Mouse) Q9QZ11
Entrez Gene ID: (Human) 9156, (Mouse) 26909
Molecular Function: exodeoxyribonuclease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support