Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: ADSLSTTKIK PLGPARASGL SKKPASIQKR KHHNAENKPG LQIKLNELWK NFGFKKDSEK LPPCKKPLSP VRDNIQLTPE AEEDIFNKPE CGRVQRAIFQ
Substrate-recognition component of the SCF (SKP1-CUL1-F-box protein)-type E3 ubiquitin ligase complex. Required for muscle atrophy following denervation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Exonuclease 1; Exonuclease I; hExo1; hExoI; rad2 nuclease family member, homolog of S. cerevisiae exonuclease 1
Gene Aliases: EXO1; EXOI; HEX1; hExoI
UniProt ID: (Human) Q9UQ84
Entrez Gene ID: (Human) 9156
Molecular Function:
exodeoxyribonuclease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support