Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MAAEAAGGKY RSTVSKSKDP SGLLISVIRT LSTSDDVEDR ENEKGRLEEA YEKCDRDLDE LIVQHYTELT TAIRTYQSIT ERITNSRNKI KQVKENLLSC KMLLHCKRD
The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and functions of exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. The complex is also essential for the biogenesis of epithelial cell surface polarity. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: augmenter of liver regeneration (ALR) pseudogene; Exocyst complex component 4; Exocyst complex component Sec8; MGC27170; REC8; rSec8; SEC8-like 1; secretory protein SEC8
Gene Aliases: ALR; C78892; EXOC4; KIAA1699; SEC8; SEC8L1; Sec8p
UniProt ID: (Human) Q96A65, (Rat) Q62824, (Mouse) O35382
Entrez Gene ID: (Human) 60412, (Rat) 116654, (Mouse) 20336
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support