Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Osenses
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Reconstitute in 500 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide is homologous with the corresponding sequence in mouse and rat.
Specificity of this antibody: SEC10.
The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and functions of exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. The complex is also essential for the biogenesis of epithelial cell surface polarity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 71 kDa component of rsec6/8 secretory complex; DKFZp666H126; Exocyst complex component 5; Exocyst complex component Sec10; hSec10; p71; p71 component of rsec6/8 secretory complex; SEC10-like 1
Gene Aliases: AI447711; AI448003; EXOC5; Gm76; HSEC10; PRO1912; SEC10; SEC10L1; SEC10P
UniProt ID: (Human) O00471, (Rat) P97878, (Mouse) Q3TPX4
Entrez Gene ID: (Human) 10640, (Rat) 60627, (Mouse) 105504
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support