Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: VFQPGVKLRD KERQIIKERF KGFNDGLEEL CKIQKAWAIP DTEQRDRIRQ AQKTIVKETY GAFLQKFGSV PFTKNPEKYI KYGVEQVGDM IDRLFDTSA
The protein encoded by this gene is a component of the exocyst complex. The exocyst complex plays a critical role in vesicular trafficking and the secretory pathway by targeting post-Golgi vesicles to the plasma membrane. The encoded protein is required for assembly of the exocyst complex and docking of the complex to the plasma membrane. The encoded protein may also play a role in pre-mRNA splicing through interactions with pre-mRNA-processing factor 19. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene, and a pseudogene of this gene is located on the long arm of chromosome 4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Exocyst complex component 7; Exocyst complex component Exo70; HGNC:23214
Gene Aliases: 2-5-3p; EX070; EXO70; Exo70p; EXOC1; EXOC7; KIAA1067; YJL085W
UniProt ID: (Human) Q9UPT5
Entrez Gene ID: (Human) 23265
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support