Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: GFSTWTYRQG YDVSIPVYSP LSAEVDLPEK GPGPRQYFLL SSQVGLHPEY REDLEALQVK HGESVLVLDK CTNLSEGVLS VRKRCHKHQV FDYPQVLQEA
This gene encodes one of two glycosyltransferases involved in the chain elongation step of heparan sulfate biosynthesis. Mutations in this gene cause the type II form of multiple exostoses. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acetylglucosaminyltransferase; Exostosin glycosyltransferase 2; Exostosin-2; Glucuronosyl-N-acetylglucosaminyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; Glucuronosyl-N-acetylglucosaminyl-proteoglycan/N; glucuronosyl-N-acetylglucosaminyl-proteoglycan/N-acetylglucosaminyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; Heparan sulfate co-polymerase subunit EXT1; Multiple exostoses protein 2; N-acetylglucosaminyl-proteoglycan 4-beta-glucuronosyltransferase; putative tumor suppressor protein EXT2
Gene Aliases: EXT2; SOTV; SSMS
UniProt ID: (Human) Q93063
Entrez Gene ID: (Human) 2132
Molecular Function: glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support